Cleanroom vacuum cleaner, EXP1-10 CR (MRAC)
Fournisseur: TIGER-VAC
110343B
TIGV110343BEA 12870 EUR InStock
TIGV110343B
Cleanroom vacuum cleaner, EXP1-10 CR (MRAC)
Aspirateurs
These wet and dry vacuum cleaner units are cleanroom compatible, autoclavable and can be used in the pharmaceutical industry. These units are also available with a middle ring with activated carbon cartridge for the adsorption of toxic/nuisance fumes and vapors.
  • All stainless steel SAE 304 construction
  • Ideal for general cleanup in hazardous locations
  • Single phase explosion proof/dust ignition proof motor
  • Less than 10 ohms of resistivity
  • Recovery tank capacity of 38 to 57 litres
  • Equipped with a true ULPA Filter (U15) located downstream of the motor
Information de livraison: Inlcudes complete static dissipating autoclavable tool kit and suction hose.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité