Industrial vacuum cleaner, AS-5 HEPA
Fournisseur: TIGER-VAC
110750B
TIGV110750BEA 764 EUR InStock
TIGV110750B
Industrial vacuum cleaner, AS-5 HEPA
Aspirateurs
This vacuum cleaner made of heavy duty plastic, is designed for the recovery of lead, asbestos, silica and other toxic/nuisance dusts.
  • 4-stage filtration system which consists of a HEPA Filter, main filter, disposable filter bag and foam exhaust filter
  • Lightweight and portable
  • Thru-Flow heavy duty motor
  • Heavy duty plastic recovery tank
  • Ready to use: filter and tool kit included
  • Mounted on a a 4-wheel dolly for easy maneuverability
This unit is equipped with a true HEPA filter (H14) locaded upstream of the motor.
Note: For dry recovery only.
Certifications: HEPA Filter - Tested: IEST RP-CC001.3. H14 by MPPS method as per EN 1822. This unit is aerosol leak tested.
Information de livraison: The unit is delivered with: suction hose (2,44 m) with plastic curved handle, extension wands (set of 2) in chrome (Ø 32 mm), curved handle in black plastic, crevice tool (Ø 32×203 mm), upholstery tool (Ø 32×130 mm), dusting brush (Ø 32×76 mm), floor tool (Ø 32×356 mm). HEPA filter (H14), cloth filter, foam exhaust filter and paper filter bags (package of 5), flat poly bags (610×610 mm) are also included.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité