Aspirateur MRV-16(4W) #110800B Wet/Dry vacuum cleaner,intercepting tank stainless steel, weight:40 Kg power: 1050 watt Scope of delivery:Plastic equipment ( suction nozzle, round tool etc.)hose, filter bags. Tank including the casters are autoclav 1 * 1 Un.
Fournisseur: TIGER-VAC
Aspirateur MRV-16(4W) #110800B Wet/Dry vacuum cleaner,intercepting tank stainless steel, weight:40 Kg power: 1050 watt Scope of delivery:Plastic equipment ( suction nozzle, round tool etc.)hose, filter bags. Tank including the casters are autoclav 1 * 1 Un.
COMMANDE
Développer tout / Réduire tout
Aspirateur MRV-16(4W) #110800B Wet/Dry vacuum cleaner,intercepting tank stainless steel, weight:40 Kg power: 1050 watt Scope of delivery:Plastic equipment ( suction nozzle, round tool etc.)hose, filter bags. Tank including the casters are autoclav 1 * 1 Un. TIGV110800B 1 * 1 Un.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou