Aspirateur AVSD-20/25L DT #111388 HEPA Filter - Included with an efficiency of 99.995% on 0.3 micron. Tested: IEST RP-CC001.3. H14 by MPPS method as per EN 1822. Pneumatically (air) operated Available with Manual Filter Shaker (MFS) For dry recovery 1 * 1 Un.
Fournisseur: TIGER-VAC
Aspirateur AVSD-20/25L DT #111388 HEPA Filter - Included with an efficiency of 99.995% on 0.3 micron. Tested: IEST RP-CC001.3. H14 by MPPS method as per EN 1822. Pneumatically (air) operated Available with Manual Filter Shaker (MFS) For dry recovery 1 * 1 Un.
COMMANDE
Développer tout / Réduire tout
Aspirateur AVSD-20/25L DT #111388 HEPA Filter - Included with an efficiency of 99.995% on 0.3 micron. Tested: IEST RP-CC001.3. H14 by MPPS method as per EN 1822. Pneumatically (air) operated Available with Manual Filter Shaker (MFS) For dry recovery 1 * 1 Un. TIGV111388 1 * 1 Un.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou