Industrial Vacuum Cleaner, C-35L EX (DT) MFS
Fournisseur: TIGER-VAC
111783B
TIGV111783BEA 0 EUR InStock
TIGV111783B
Industrial Vacuum Cleaner, C-35L EX (DT) MFS
Aspirateurs
Industrial vacuum cleaner for continuous duty. Designed specifically for housekeeping and general cleaning. It is ideal for general cleaning and for process integration.
  • Static dissipating
  • HEPA/SD accessories - included
  • High filtration efficiency unit
  • Stainless general
  • Dry recovery and detachable tank
  • Brushless motor available
  • By-pass Ametek® long life motor
Longer life brushless motor available. Always with 4×H14 HEPA filters, Manual Filter Shaker (MFS) with static dissipative teflon laminated main cloth filter assembly, Detachable recovery tank (DT) for easy disposal of recovered materials, Material to be recovered within a conductive poly liner recovery bag for easy disposal.
Information de livraison: Includes 4 HEPA filters. Class H14. Individually factory tested.
Attention: Note: For dry recovery only.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité