Vacuum Cleaner, 2D-10 (4W) HEPA ORDLOC
Fournisseur: TIGER-VAC
113185B_BLSD
TIGV113185B_BLSDEA 0 EUR InStock
TIGV113185B_BLSD
Vacuum Cleaner, 2D-10 (4W) HEPA ORDLOC
Aspirateurs
Ideal for housekeeping and general cleaning. For the safe collection of combustible dusts in ordinary locations/areas and unclassified zones.
  • Ignition source safe/protected
  • Protected against ingress of dust IP5X
  • Very high level of protection inside the collection tank
  • Heavy duty two-stage by-pass motor
  • Longer life brushless motor available. Always with 2× H14 HEPA filters
  • Filter blockage indicator light
HEPA filter with an efficiency of 99,995% at 0,3 µm. Tested IEST-RP-CC001. H14 by MPPS method as per EN 1822 and OSHA compliant. All of our HEPA vacuum systems are aerosol leak tested before leaving our facility - included.
A package of the disposable conductive 'wet' recovery bags is included with the vacuum cleaner. Stainless steel tank Type 430 fits any application.
Cart type: Four wheel dolly (4W)
Type (powerhead): Electric
Dry recovery - disposable filter bag: 19 L
Tension: 220 - 240 V
Certifications: Meets EU Safety, Health and Environmental Protection Requirements.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité