Size exclusion chromatography columns, TSKgel® UP-SW Aggregate
Fournisseur: TOSOH Bioscience
0023524 0023525 0023526 0023527
TOYO0023524EA 3090 EUR InStock
TOYO0023524 TOYO0023525 TOYO0023526 TOYO0023527
Size exclusion chromatography columns, TSKgel® UP-SW Aggregate
Colonnes pour chromatographie
TSKgel® UP-SW Aggregate, the latest addition to the UP-SW series of silica-based size exclusion chromatography (SEC) columns, are specifically designed to facilitate the analysis of mAb aggregates, high molecular weight (MW) proteins and nulceic acids.
  • Optimized for large proteins and higher order mAb aggregates
  • Proven TSKgel® SW SEC quality
  • Provides high sensitivity and excellent column efficiency
  • Compatible with both HPLC and UHPLC systems
  • Calibration range (proteins):10 - 2000 kDa
These columns are 3 µm, 30 nm pore size SEC analytical columns, that have been designed with a higher exclusion limit than other TSKgel® UP-SW columns.
Available in 4.6 mm ID, TSKgel® UP-SW Aggregate columns are compatible with both UHPLC and HPLC systems and require less sample while delivering higher sensitivity compared to the 7.8 mm ID TSKgel® UltraSW Aggregate columns.
These columns feature high pore volume, low sample adsorption (due to the derivatization of the particle surface with ligands containing diol functional groups) and excellent column efficiency, all contributing to unsurpassed sample resolution.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité