FcR-IIIA-5PW, TSKgel®
Fournisseur: TOSOH Bioscience
0023532
TOYO0023532EA 2580 EUR InStock
TOYO0023532
FcR-IIIA-5PW, TSKgel®
Colonnes pour chromatographie
TSKgel FcR-IIIA-5PW is a semi-preparative affinity column which immobilises the recombinant FcγRIIIA ligand bonded to pourous 10 µm polymethacrylate particles which can load up to 5 mg of mAb.
  • Semi-preparative affinity column for collecting fractions with different FcR affinities to characterise them
  • Temperature range 15 to 25 °C
Isolate antibodies according to FcR Affinity from a heterogeneous sample. Expression of antibodies typically leads to an antibody mixture with different affinities to the FcγIIIa receptor. The TSKgel FcR-IIIA-5PW column can be loaded with 5 mg of antibody sample and resolves three affinity fractions. These can be employed to analyse structural and functional differences linked to differences in FcR affinity.
Information de livraison: Shipping solvent: 0,65 mmol citric acid, 9,35 mmol/L trisodium citrate containing 0,025 ProClin® 300, pH 6,5.
Conditionnement: Each package includes the related HPLC column as well as the operating conditions and specification sheet.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité