Detergent, heavy duty, floor detergent, Opal
Fournisseur: TRISTEL SOLUTIONS
CRY313
TRSTCRY313EA 0 EUR InStock
TRSTCRY313
Detergent, heavy duty, floor detergent, Opal
Produits nettoyants Détergents et Nettoyants de Surfaces
A non foaming, heavy duty, floor detergent to be used manually or through floor scrubbing machines. Opal is excellent at removing organic soils, including fats and oils, and is safe for use on most floor surfaces including vinyl, stone and concrete.
  • Heavy duty detergent for use on organic soils
  • Suitable for manual use or use in floor scrubbing machines
  • Dilution for use: between 1 to 2% v/v
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
Information de livraison: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
Conditionnement: Available in 5 litre pack size only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité