Face shield visor, 701.02
Fournisseur: UNIVET S.R.L.
1220010
UNVT1220010EA 106 EUR InStock
UNVT1220010
Face shield visor, 701.02
Écrans de protection
The 701.02 face shield meets rigorous EN 166 standards (droplets and splashes) by providing visor with extended coverage on the upper part of the forehead.
  • Anti-fog acetate visor
  • Adjustable elastic head band
  • Visor inclines up to 90°
  • Interchangeable visor
  • Face shield suitable for use in combination with prescription glasses and protective masks
This anti-fog visor allows maximum vision due to anti-fog characteristics. A soft sponge in front with an easily adjustable band ensures comfort. This visor is suitable for use with corrective glasses and respiratory protective masks.
Certifications: Certified according to EN 166, marking 3 protection against droplets and liquid splash.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité