illustra™ ExoProStar™ S kits, Cytiva
Fournisseur: Cytiva
US79050
US79050EA 341 EUR InStock
US79050
illustra™ ExoProStar™ S kits, Cytiva
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
The ExoProStar™ S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar™ S improves digestion efficiency with no degradation of the target PCR product. Sample remain completely intact and ready for immediate use in manual or automated processes. For maximum flexibility the enzymes are supplied in two separate tubes.
  • Enzymes optimised to work together for high efficiency removal of unincorporated primers and nucleotides
  • Enzymes provided in two separate tubes - just two simple pipetting steps are needed to prepare the reaction
  • Fast 15 minute protocol
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
  • Complete heat inactivation of the enzymes within 10 minutes
The ExoProStar S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar S improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité