Cell Culture Media, Ames' Media
Fournisseur: US Biological
A1372-25A-100L
USBIA1372-25A-100LEA 843 EUR InStock
USBIA1372-25A-100L
Cell Culture Media, Ames' Media
Milieux de culture cellulaire
Ames' medium was formulated to support retinal tissue in relatively short-term culture.
  • White to pale yellow powder
  • Homogeneous, free flowing powder
Rabbit retina has been incubated in Ames' medium for over 2 days with its metabolism and electrical responses to light stimuli well maintained. This mixture is a medium of choice for maintaining central nervous system tissue in vitro.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité