(±)-O-Desmethylvenlafaxine-D6 solution, certified reference material, Cerilliant®, Supelco®
Fournisseur: Merck
V-027-1ML
V-027-1MLEA 245 EUR InStock
V-027-1ML
(±)-O-Desmethylvenlafaxine-D6 solution, certified reference material, Cerilliant®, Supelco®
Étalons Étalons organiques
A certified reference solution suitable for use as an internal standard for quantitation of O-desmethylvenlafaxine levels in urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis.
Also known as desvenlafaxine, O-desmethylvenlafaxine is an SNRI antidepressant marketed under the trade name Pristiq® for treatment of depression. O-desmethylvenlafaxine is also a major urinary metabolite of venlafaxine, an SNRI antidepressant sold as Effexor® or Efexor and used to treat major depressive disorder, generalised anxiety disorder, and other anxiety disorders associated with depression.
Conditionnement: Snap-N-Spike®
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37f1a268282117466b5649d0ada92afdd2e9932a&vid=37f1a268282117466b5649d0ada92afdd2e9932a&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité