Rotary Vane Pump, RE 9
Fournisseur: VACUUBRAND
20797170
VACU20797170EA 3265 EUR InStock
VACU20797170
Rotary Vane Pump, RE 9
Pompe Pompes à vide à palettes
Rotary vane pumps from VACUUBRAND offer great performance in a compact design. Even with the gas ballast open, they enable a very good ultimate vacuum. You also benefit from extended oil change and maintenance intervals thanks to optimized lubrication circuit and large usable oil volume.
  • Very high flow rates even at vacuum levels approaching ultimate vacuum
  • High water vapor tolerance due to efficient gas ballast
  • Very good ultimate vacuum even with gas ballast
  • Vacuum-tight at switch-off
  • External anti-suckback valve not needed
  • Large oil volume: Long intervals between oil changes
  • Ease of maintenance due to telescopic design
The RE 9 is a single-stage powerful rotary vane pump for fast evacuation of vessels and at higher gas flow rates. Typical applications are freeze drying, fine vacuum distillation and drying ovens. The single-stage rotary vane pump RE 9 is also available in various packages with matching accessories such as exhaust filter FO and valve VS 25. Rotary vane pumps are used for a process vacuum of a few mbar to 10⁻³ mbar. 40 mbar water vapor tolerance.
Information de livraison: Pump oil filled and completely mounted, ready for use, with manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité