Merlin Microseal
Fournisseur: VARIAN
392609901
VARI392609901EA 1250 EUR InStock
VARI392609901
Merlin Microseal
Matériel de chromatographie Chromatographie, Pièces de rechange
The Merlin Microseal is available in standard and high pressure kits.
  • Low bleed, longer life alternative to standard septa for split/splitless injection and SPME
  • Requires 23 gauge syringe needle
  • Has a lifetime of more than 2000 injections, depending on samples and operating conditions
  • Greatly reduced instrument downtime for septa changes and injection port liner changes due to septa particulates
  • Two distinct sealing mechanisms: double O-ring type seal around the syringe needle and spring assisted duckbill to seal the injection port
  • Temperature range 50 to 400 °C
During normal use, when the autosampler syringe needle tip is wiped clean by the sample vial septum, no solvent comes in contact with the Microseal septum. Traces of solvent which might contact the septum evaporate rapidly. However, exposure to liquid ethers and similar oxygenated solvents causes deterioration.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité