Colonnes capillaires, biodiesel sélectionné
Fournisseur: VARIAN
CP6530 CP9076 CP9077 CP9078 CP9079 CP9080 CP9083
VARICP6530EA 119 EUR InStock
VARICP6530 VARICP9076 VARICP9077 VARICP9078 VARICP9079 VARICP9080 VARICP9083
Colonnes capillaires, biodiesel sélectionné
Colonnes pour chromatographie
Select Biodiesel columns address the key challenge of good column lifetime when operating at very high temperatures up to 400 °C.
  • Complete set of biodiesel columns for full compliance and ease-of-use
  • UltiMetal technology provides high accuracy and longevity
  • Designed and pre-tested for complete confidence in results
  • Results are more consistent
  • No column breakage
Select Biodiesel columns are offered with a pre-coupled retention gap that is leak tested prior to shipment, making life much easier for the operator. This short piece of tubing not only enhances the analytical separation but also dramatically simplifies automation when using the column with a column inlet as specified in the standard methods.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité