Gel permeation chromatography columns, MesoPore™
Fournisseur: VARIAN
PL1113-1325 PL1113-6325
VARIPL1113-1325EA 590 EUR InStock
VARIPL1113-1325 VARIPL1113-6325
Gel permeation chromatography columns, MesoPore™
Colonnes pour chromatographie
MesoPore columns have been specifically designed to give optimum results in the analysis of prepolymers, i.e. polymeric materials with a large oligomeric component.
  • Full solvent compatibility with no detrimental effect on efficiency
  • Low particle size extracts maximum information from the analyte
  • No MWD dislocations so the distribution is a faithful representation of the sample
By combining a 3 µm particle size with high pore volume, MesoPore columns give the highest resolution separations for the analysis of low molecular weight polymers, such as prepolymers, resins, polyols and siloxanes.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité