Gel Permeation Chromatography Columns, PL HFIPgel
Fournisseur: VARIAN
PL11146900HFIP
VARIPL11146900HFIPEA 2420 EUR InStock
VARIPL11146900HFIP
Gel Permeation Chromatography Columns, PL HFIPgel
Colonnes pour chromatographie
Hexafluoroisopropanol (HFIP) is used as a solvent in GPC for the analysis of important industrial polymers such as polyesters, polyamides and polylactide/glycolide copolymers. For greatly improved performance in extremely polar solvents such as HFIP and trifluoroethanol, we have developed novel 'multipore' technology to produce PL HFIPgel, a PS/DVB packing featuring a monodisperse particle size, high pore volume and high resolution.
  • Optimized separation range delivers high performance with no artifacts
  • Low operating pressure reduces system wear and unnecessary downtimes
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité