SPME fibers
Fournisseur: VARIAN
SU57326U
VARISU57326UEA 0 EUR InStock
VARISU57326U
SPME fibers
Matériel de chromatographie Chromatographie, Pièces de rechange
SPME fibers are available in a range of chemistries, formats, and for use with autosamplers or manual injections.
  • SPME fibers can be used multiple times when treated with the proper care and caution
  • Each fiber has a colour-coded or notched hub, indicating the type of coating on the fiber
Kits are also available to support method development.
Informations de commande: When ordering SPME fibers, note that the fiber kits contain fibers only. For a first-time order, you will also need to order the appropriate fiber holder for your needs.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité