Cleanroom wipes, pre-saturated, DAS-WIPE 100 Sterile, VAI®
Fournisseur: VELTEK ASSOCIATES
DW1001212S2319
VELTDW1001212S2319EA 1230 EUR InStock
VELTDW1001212S2319
Cleanroom wipes, pre-saturated, DAS-WIPE 100 Sterile, VAI®
Lingettes Lingettes pour salles propres Lingettes pour salles propres, présaturées
Individually-packaged sterile wipes are a polyester blend that is inherently low in particulate and shedding features and saturated with DAS Stainless Steel Cleaner and Lubricant. DAS-Wipe 100 is designed to replace silicone on turntables, process lines and as a general lubricant to assure bottles do not stick during movement towards the critical fill site.
  • Laundered and packaged in ISO 4 conditions, closed filling in an ISO 5 Cleanroom (Grade A)
  • 100% synthetic filament constructed in a continuous knit that minimizes fiber and particulate release
  • Formulated with WFI
  • Filtered at 0,2 µm
  • Cut using FocusEdge heat cutting technology
  • Gamma-irradiated, double-bagged
Excellent for pharmaceutical and biotechnology industries. Delivered with lot-specific sterile documentation. Completely traceable. Shelf life of three years.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité