Wipedown HC dry wipes, VAI®
Fournisseur: VELTEK ASSOCIATES
VEL13-9X9S3013
VELTVEL13-9X9S3013EA 156 EUR InStock
VELTVEL13-9X9S3013
Wipedown HC dry wipes, VAI®
Lingettes Lingettes pour salles propres Lingettes pour salles propres, stériles
WipeDown HC dry wipers are ultra-clean laundered and suitable for use in cleanroom environments.
  • Sterile versions are sterility assured through gamma irradiation at a 10⁻⁶ SAL
  • Individually labelled with lot number and expiration date
  • Packaged neatly folded for easy removal
  • Completely lot traceable
  • Delivered with lot specific documentation
  • High absorbency and low in particulate shedding and soluble extracts
  • Minimises fibre and particulate release
  • Validated for sterility and shelf life
Constructed of 55% polyester and 45% cellulose spunlace blend. Particulates, residues, and foreign particles are effectively entrapped in the webbing of both of these materials.
Uses a proprietary cutting method, FocusEdge heat cutting technology. This ensures that there is a reduction in edge particulates on the wiper that is typically left behind after laser cutting.
WipeDown HC dry wipers can be used for filling and packaging machinery wipe downs, IPA wipe downs, spill control, general wiping, and contamination control.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité