Ultra Chemical Resistant Tubing for Tygon® Ink 1000
Fournisseur: Saint Gobain Life Sciences
A0C1S1520
VERNA0C1S1520EA 233 EUR InStock
VERNA0C1S1520
Ultra Chemical Resistant Tubing for Tygon® Ink 1000
Tuyaux Transfer Tubing
For UV inks in particular, UV blocking properties is an industry standard. Tygon® Ink 1000 has shown over an accelerated testing period that it can maintain the ink integrity with our patented technology, making it an ideal candidate for the UV market.
  • Outstanding chemical resistance to small acrylic monomers
  • Flexibility
  • UV blocking
  • Low extractable levels
  • Long service life
Chemical Resistance: In the ink transfer space, chemical compatibility and flexibility are two of the most important criteria for ensuring satisfactory performance. Tygon® Ink 1000 was specially engineered to deliver premium performance in an environment where UV inks are used such as digital printing and 3D printing applications. This product has been tested with a variety of diluents (IBOA, VMOX, HDDA, etc.) and has shown excellent chemical resistance properties over an extended testing phase.
Typical Applications: Digital printing, inkjet printing, wide format printing, packaging printing, 3D printing.
Vacuum rating: 23 °C (mmHg)
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité