Boiling stones
Fournisseur: Saint Gobain Life Sciences
D1069103
VERND1069103EA 47 EUR InStock
VERND1069103
Boiling stones
Pierres d'ébullition
Ultra-pure, virgin PTFE boiling stones promote gentle, efficient boiling and help prevent overheating, even of highly volatile liquids. Ideal for continous extraction methods. Stones minimise 'bumping' during reflux and distillation.
  • Chemical resistant
  • Ideal for boiling organic liquids
  • Minimises superheating
The boiling stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis. They are easily cleaned and non-contaminating. Shows no trace of face acidity or alkalinity. They will not scratch or abrade glassware and leave noMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité