Sani-Tech® SPT-60L Tubings, Silicone, Platinum Cured
Fournisseur: Saint Gobain Life Sciences
SPT60L-236-394
VERNSPT60L-236-394EA 379 EUR InStock
VERNSPT60L-236-394
Sani-Tech® SPT-60L Tubings, Silicone, Platinum Cured
Tuyaux Transfer Tubing
Sani-Tech® SPT-60L is a premium platinum-cured silicone tubing designed for the most exacting pharma operations where low spallation, long pump life and high accuracy are required. Sani-Tech® SPT-60L provides best-in-class accuracy and precision assuring consistent filling performance and reducing the potential for costly over/under fills.
  • Filling accuracy ((dispensing target fill volume) and Precision (repeatability of delivered volume)
  • Biocompatible
With an average pump life over 250 hours, Sani-Tech® SPT-60L tubing outperforms extruded silicone tubing providing improved performance and reliability under various pumping conditions. Sani-Tech® SPT-60L has the lowest spallation (number of particles and area of particles) across multiple dispensing cycles among the various silicone pump tubing evaluated.
Certifications: ISO 10993-3 / ISO 10993-5, ADCF/BSE/TSE statements, USP <88> Class VI, EP 3.1.9.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité