Sani-Tech® SPT-60L Tubings, Silicone, Platinum Cured
Fournisseur: Saint Gobain Life Sciences
SPT60L-236-394
VERNSPT60L-236-394EA 0 EUR InStock
VERNSPT60L-236-394
Sani-Tech® SPT-60L Tubings, Silicone, Platinum Cured
Tuyaux Transfer Tubing
Sani-Tech® SPT-60L is a premium platinum-cured silicone tubing designed for the most exacting pharma operations where low spallation, long pump life and high accuracy are required. Sani-Tech® SPT-60L provides best-in-class accuracy and precision assuring consistent filling performance and reducing the potential for costly over/under fills.
  • Filling accuracy ((dispensing target fill volume) and Precision (repeatability of delivered volume)
  • Biocompatible
With an average pump life over 250 hours, Sani-Tech® SPT-60L tubing outperforms extruded siliconeMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité