Tubing, silicone, platinum cured, Sani-Tech®, STHT–C series
Fournisseur: Saint Gobain Life Sciences
STHT-C-125-3H
VERNSTHT-C-125-3HEA 585 EUR InStock
VERNSTHT-C-125-3H
Tubing, silicone, platinum cured, Sani-Tech®, STHT–C series
Tuyaux Transfer Tubing
STHT®-C silicone tubing is an ultra-pure, biopharmaceutical-grade tubing which resists temperature extremes, ozone, corona, radiation, moisture, compression sets, weathering and chemical attack and imparts no taste or odours to fluids transported within it.
  • Unsurpassed surface smoothness
  • Ultra-flexible
  • Temperature resistance from –62 to 260 °C
Permanently laser marked with part number and lot code for total product traceability. Used in sterile fill and transfers, drug delivery, media processing, sanitary peristaltic pump tubing, laboratory use, cell harvest and liquid chromatography.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité