Mops, SuperMop AG and SuperMop Micro AG
Fournisseur: FHP VILEDA SCS
100273
VILE100273EA 0 EUR InStock
VILE100273
Mops, SuperMop AG and SuperMop Micro AG
Serpillières
SuperMop Ag and SuperMop Micro Ag are the top hygienic solutions for fast, effective and intensive cleaning of small and medium sized floor areas. The unique construction that hinders the growth and multiplication of bacteria, now has silver ions added that inhibits 99,99% of bacteria growth within the mop.
  • Independently tested to inhibit bacteria growth
  • Oval design, allows easy cleaning of corners
  • High resistance to detergents and bleaches
  • Low friction, lightweight non-woven-material
  • Highly absorbent for spill pick up
  • Colour-coded in four colours to avoid cross contamination
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité