Stereo microscope, digital, 3-D full high definition with zoom, Ergonomic, DRV-Z1
Fournisseur: VISION ENGINEERING
DRVZ502 DRV001 DRV250 DRV233
630-3461EA 21624 EUR InStock
630-3461 VISIDRV001 VISIDRV250 VISIDRV233
Stereo microscope, digital, 3-D full high definition with zoom, Ergonomic, DRV-Z1
Microscopes
Specifically designed for inspection and manufacturing applications, DRV-Z1 is a digital stereo 3-D viewer with zoom that brings together the benefits of optical stereo microscopy and digital technology into a single unique system.
  • Digital stereo 3-D with vivid depth perception
  • 4 million pixel image
  • Precision hand-to-eye co-ordination
  • Improved comfort and productivity
  • New real time collaboration opportunity
For users, DRV-Z1's stereo digital image provides a natural 3-D view, with full high definition (FHD) resolution and excellent subject clarity,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité