High definition digital microscope, EVO Cam II
Fournisseur: VISION ENGINEERING
ECL200 ECL003 ECL150 ECL001 ECL002
VISIECL200EA 1066 EUR InStock
VISIECL200 VISIECL003 VISIECL150 VISIECL001 VISIECL002
High definition digital microscope, EVO Cam II
Microscopes
Making image capture and documentation simpler, faster and more reliable - wherever ultra-sharp images are required for inspection, fault analysis and much more.
  • Exceptional high resolution 1080 p / 60 fps image quality
  • Intuitive easy image capture and documentation
  • Stand-alone, wireless or PC connectivity
  • High quality stands for precision and stability
  • Unique 360° rotating viewer
  • Ultra-sharp image quality with full autofocus
  • Optical magnification range 1,7 to 300×
  • Maximum magnification with digital zoom 3600×
  • Fast 60 fps provides clear images without
More Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité