Ergonomic Eyepiece-less Stereo Microscope with Verso Articulated Arm, Mantis Ergo
Fournisseur: VISION ENGINEERING
SMR201
VISISMR201EA 4254 EUR InStock
VISISMR201
Ergonomic Eyepiece-less Stereo Microscope with Verso Articulated Arm, Mantis Ergo
Microscopes
Mantis Ergo compact ergonomic stereo microscope. Easy to use with interchangeable lenses. Flexible articulated arm. Magnification from 3 to 15× (objectives to be added).
  • High quality optical stereo image
  • Ergonomic working position
  • Long working distance
  • 3 lens multi-view turret
  • Dynamic 3D lighting
  • White/UV lighting option
Eyepiece-less design. Mantis eyepiece-less technology projects a large high quality 3D image directly to your eye, making it easier to see and more comfortable to view than a binocular stereo microscopes. The clarity of the 3D image supports enhanced hand-eye coordination, making subject manipulation with tools more accurate, faster and more efficient. You can even wear corrective or protective eyewear without loss in performance.
Compatible objectives: 3×, 4×, 6×, 8×, 10×, 15×, 6× SLWD, 8× SLWD.
Certifications: CE, ISO 9001.
Informations de commande: Requires at least one objective to choose in the accessories list.
Information de livraison: Supplied with a user manual and international power supply.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité