Ergonomic Eyepiece-less Stereo Microscope with Stabila Bench Stand and Substage Illumination, Mantis Ergo
Fournisseur: VISION ENGINEERING
SMR212
VISISMR212EA 4823 EUR InStock
VISISMR212
Ergonomic Eyepiece-less Stereo Microscope with Stabila Bench Stand and Substage Illumination, Mantis Ergo
Microscopes
Mantis Ergo compact ergonomic stereo microscope. Easy to use with interchangeable lenses. Bench stand with integrated substage illumination. Magnification from 3 to 15× (objectives to be added).
  • High quality optical stereo image
  • Ergonomic working position
  • Long working distance
  • Three lens multi-view turret
  • Dynamic 3D lighting
  • White/UV lighting option
Eyepiece-less design. Mantis eyepiece-less technology projects a large high quality 3D image directly to your eye, making it easier to see and more comfortable to view than a binocular stereo microscopes. The clarity of the 3D image supports enhanced hand-eye coordination, making subject manipulation with tools more accurate, faster and more efficient. You can even wear corrective or protective eyewear without loss in performance.
Compatible objectives: 3×, 4×, 6×, 8×, 10×, 15×, 6× SLWD, 8× SLWD.
Certifications: CE, ISO 9001.
Informations de commande: Requires at least one objective to choose in the accessories list.
Information de livraison: Supplied with a user manual and international power supply.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité