Repetitive pipettes, macro, electronic, Ovation®
Fournisseur: VISTALAB TECHNOLOGIES
1065-10ML
VIST1065-10MLEA 1420 EUR InStock
VIST1065-10ML
Repetitive pipettes, macro, electronic, Ovation®
Pipettes Pipettes à répétition
These macro pipettes with eject button are fully electronic, providing a broad menu of liquid handling functions.
  • Suitable tips offer easy access into tubes, bottles and cell culture flasks
  • For rapid, precise and accurate dispensing, up to 50 times from a single aspiration
  • Innovative design provides relaxed grip and comfortable posture while pipetting
  • Pipettes stand on their own
Volumes, speeds and repetition settings are easy to setup, store and instantly recall for pipetting, mixing, serial diluting or repeat dispensing. When extra length is neededMore Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité