Antimousse silicone GAMMA EGT SAG 471 pour la production
997002.6002EA 630 EUR InStock
997002.6002
Antimousse silicone GAMMA EGT SAG 471 pour la production
Antimousse silicone
Silicone antifoam liquid , sterilised by gamma irradiation. SAG 471 antifoam is for use in both aqueous and non-aqueous systems, imparting outstanding durability in the control of foam over time. Highly effective (even at low concentrations), it offers cost-efficient foam control for a variety of applications, including aerobic fermentation of antibiotics involving actinomycetales organisms, glycol-based coolants, heat exchange systems, adhesives, sealants, vaccines, and others. SAG 471 antifoam is self-dispersing in water.
  • Maximization of fermentor capacities
  • Excellent durability
  • Excellent foam control performance in fermentation processes
  • Self-dispersing in water- suitability under sterilization
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité