640490
VWRK640490EA
73.1
EUR
InStock
VWRK640490
Hématoxyline de fer solution 1 Weigert's, Clin-Tech
Iron hematoxylin
Weigert's solutions are used in histology for the coloration and detection of elastic fibers in connective tissues.
- Weigert’s solution 1 – 1% haematoxyline in alcohol
- Weigert's solution 2 – 1,2% iron chloride in 1% hydrochloric acid solution