VWR®, Davidson's fixative
Fournisseur: KLINIPATH (PL)
641139
VWRK641139EA 154 EUR InStock
VWRK641139
VWR®, Davidson's fixative
Colorants Réactifs pour la microscopie et la coloration
Davidson's (sometimes called Hartmann's) fixative is a rapid fixative which gives good nuclear detail with minimal formalin pigment. Specimens cut and stain as if fixed in ordinary formalin.
  • Fixation of small samples is rapid
  • Contains no mercury or other metals
Davidson's fixative is particularly useful for preparing tumours, bone marrow specimens, gynecologic material, fatty breast, and medical biopsies. It may be used for testicular biopsies and for overnight fixation of fat containing lymph nodes.
Attention: Exposure to the fixative should be limited to 24 hours (tissue may be transferred to neutral buffered formalin or 70% alcohol for storage).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité