VWR®, Microcentrifuge tubes, with caps, standard line
Fournisseur: VWR Collection
10025-726
VWRU10025-726EA 72.7 EUR InStock
VWRU10025-726
VWR®, Microcentrifuge tubes, with caps, standard line
Tubes Microtubes à centrifuger
These microcentrifuge tubes with caps are made of super-clear, high quality USP Class 6 gamma resistent PP.
  • Available in three volumes: 0,5, 1,5, and 2,0 ml
  • Engraved graduations ensure accuracy
  • Flat and frosted caps surface together with smooth and frosted body surface
  • Clear and legible marking area provided
  • Tubes can be spun up to 25000 ×g
  • Autoclavable at 121 °C and freezable to −80 °C
  • DNase-free, RNase-free and non pyrogenic
They feature a positive seal design that withstands repeated cap closures and will not pop open during boiling. Tubes fit all standard rotors.
Certifications: TSE/BSE-free certified.
Conditionnement: Bagged tubes are packed in durable and re-sealable (zip closure) bags. Inner packaging is individually labelled for lot-to-lot traceability.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité