VWR® CryoGuard, Cryo Gloves
Fournisseur: VWR Collection
97008-246
VWRU97008-246EA 167 EUR InStock
VWRU97008-246
VWR® CryoGuard, Cryo Gloves
Gants
These multi-layer, insulated gloves are designed to protect hands and arms from the hazards of working with cryogenic materials. They can be worn for extended periods of time, as they are extremely warm, even in temperatures as low as –196 °C.
  • Lightweight, flexible, and durable
  • Inner lining wicks moisture from the skin
  • All wrist-length gloves also incorporate an uninsulated knit cuff
Water-resistant and waterproof gloves feature two layers of insulation material bonded together at the edges to trap insulating air without adding additional bulk and an inner lining to wick away moisture from the skin. Waterproof gloves incorporate a thin, seamless, 100% waterproof inner protective layer.
Attention: Gloves are not intended for immersion in liquid nitrogen.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité