Limulus PS Single Test
Fournisseur: FUJIFILM WAKO CHEMICALS
299-54501
WAKF299-54501EA 1710 EUR InStock
WAKF299-54501
Limulus PS Single Test
Tests microbiologiques Tests sur les endoxines
The Limulus PS Single Test is composed of LAL endotoxin-specific single-test vials and an affinity resin suspension called PyroSep™, designed to overcome any product interference by adsorbing potential endotoxin in samples while washing away the inhibitory components.
  • Endotoxin-specific reagent avoids false positive results from glucans
  • This kit is in single test configuration to avoid contamination
  • KTA quantitation for this product is performed on Toxinometer®
  • Allows endotoxin-specific measurement in samples that typically have inhibitory components: ethanol, silicone oil, soybean oil, olive oil, stearic acid, egg yolk lecithin, oil-adjuvanted vaccine, albumin products, coagulation factor products (factor 8), interferon formulation, antithrombin III, immunoglobulin product and other ethanol-soluble samples
  • Measurement system: Toxinometer
When using this product, the endotoxin in the sample is adsorbed onto a capillary column filled with PyroSep™ (an affinity adsorbent with histidine bound to a water-insoluble support via a spacer). Inhibitory substances contained in the sample are washed away, allowing the adsorbed endotoxin to be measured with the single-test format LAL reagent.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité