Biochrom Biowave 3+
Fournisseur: Biochrom
80-3007-37
WAPR80-3007-37EA 8080 EUR InStock
WAPR80-3007-37
Biochrom Biowave 3+
Spectrophotomètres et Photomètres Spectrophotomètres UV-Visible
The Biowave 3 series offers all the benefits and capabilities of a research grade Life Science spectrophotometer with robust design and reasonable cost that also makes it perfect for students.
  • Split beam (RBC) UV/Vis spectrophotometer 190 to 1100 nm (3/5 nm bandwidth)
  • Pre-programmed methods for DNA, RNA, oligos, proteins and cell density
  • Nucleic acid scans for purity checking
  • Low volume capability with microvolume cuvettes
  • Novel optics with a xenon source for long lamp lifetime and low running costs
  • Lightweight and stackable for storage
  • Comprehensive IQ/OQ documentation and service support is available for our Biowave instruments
Results may be downloaded to a USB flash drive or printed to an optional integrated graphical printer for a permanent record. Alternatively, results can be exported via a cable connection to a suitable PC running Print Via Computer (PVC) software or wireless accessory (optional).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité