Biochrom Biowave 3+ Colour Touch
Fournisseur: Biochrom
80-3007-57
WAPR80-3007-57EA 0 EUR InStock
WAPR80-3007-57
Biochrom Biowave 3+ Colour Touch
Spectrophotomètres et Photomètres Spectrophotomètres UV-Visible
The Biowave 3 series offers all the benefits and capabilities of a research grade Life Science spectrophotometer with robust design and reasonable cost that also makes it perfect for students.
  • The Biowave 3+ Color Touch is truly an all-encompassing life sciences spectrophotometer
  • From the most basic cell density or single wavelength reading, to low volume molecular quantification, this is the most versatile single beam UV/Visible instrument on the market
  • This compact and robust instrument is perfect for most Life Science laboratories, from undergraduate
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité