Colorimeter, Libra S2
Fournisseur: Biochrom
80-5000-02
WAPR80-5000-02EA 1500 EUR InStock
WAPR80-5000-02
Colorimeter, Libra S2
Colorimètres Colorimètres multiparamètres
The Libra S2 is a small, robust, easy to use instrument that has been designed with both the student and field user in mind. It is ideal for teaching principles of science and analysis in schools and colleges.
  • Rechargeable NiMH batteries
  • Extremely versatile and ideal for QC and educational applications
  • Instrument surface is designed for ease of cleaning and decontamination
The instrument measures in absorbance and % transmission mode as well as in simple kinetics, enabling changes in absorbance over time and reaction rates to be determined. The instrument may be linked via a serial lead to either a serial printer for hardcopy output or to a PC for download of results to spreadsheet. It has an analogue output, and may also be connected to a chart recorder using a standard 2×4 mm socket to output absorbance-time data when in kinetics mode.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité