Ion exchange papers, grade SG81, Whatman™
Fournisseur: Whatman products (Cytiva)
3668-915
WHAT3668-915EA 238 EUR InStock
WHAT3668-915
Ion exchange papers, grade SG81, Whatman™
Filtres Papiers filtre Papier pour chromatographie
These ion exchange papers combine cellulose and large pore silica gel for separations in which both partition and adsorption are important, including the separation of phospholipids, steroids, phenols and dyes.
  • Simultaneous development of multiple samples on the same sheet
  • Sequential development of the same samples with different solvents and/or different concentrations of the same solvent
  • Suitable for two-dimensional chromatography (change in direction of the solvent front) with possible improved resolution
  • Used in enzyme assays to separate product from
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité