Deep well plate, 96-well, WHEATON® μLplate®
Fournisseur: DWK Life Sciences
07-2000
WHEA07-2000EA 15.5 EUR InStock
WHEA07-2000
Deep well plate, 96-well, WHEATON® μLplate®
Microplaques Microplaques deepwell
500 μl round well, round bottom, 96-well microplate.
This 96-well round well microplate is made of polypropylene and available in a variety of volumes. Microplates are manufactured to ANSI specifications to guarantee a smooth integration into the automated work flow. Round and square well covers are available separately as pierceable, standard, and pre-cut. Covers manufactured of Ethylene Vinyl Acetate (EVA) are pierceable by standard needle configurations and have a superior seal to similar products. For those needing reseal ability, the available silicone mats with a spayed-on PFTE barrier are available as standard or pre-cut.
Certifications: ANSI/SLAS 1-2004 through ANSI/SLAS 4-2004.
Informations de commande: Cover not included.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité