WHEATON® Filtration Assembly with Stainless Steel Support and Standard Taper 40/35 Joint Connection, 47 mm
Fournisseur: DWK Life Sciences
419390
WHEA419390EA 658 EUR InStock
WHEA419390
WHEATON® Filtration Assembly with Stainless Steel Support and Standard Taper 40/35 Joint Connection, 47 mm
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants en verre
This filtration assembly are designed for routine filtration of organic solvents, corrosive liquids and the removal of particulates from HPLC solvents.
  • Glass components are manufactured from WHEATON 33 low extractable borosilicate glass
  • Ground glass connection eliminates phthalate contamination that can occur from silicone or neoprene stoppers
  • Drip tip on the support bases extends below the hose connection preventing filtrate from entering the vacuum line
  • Recommended for filtering viscous or proteinaceous solutions or to produce ultraclean filtrate
Units have a #2 hose connection for 1/4” (6 mm) I.D. flexible tubing.
Certifications: Glass conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Information de livraison: Includes a 316 stainless steel support with 120 mesh screen, PTFE support screen gasket, glass support base, 300 ml graduated funnel, anodized aluminium clamp and 1 L filter flask.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité