Vacuum filtration complete units, WHEATON®
Fournisseur: DWK Life Sciences
419450 419462
WHEA419450EA 1120 EUR InStock
WHEA419450 WHEA419462
Vacuum filtration complete units, WHEATON®
Entonnoirs Entonnoirs filtrants avec membrane
Borosilicate glass, with stopper or standard ground joint connection.
  • Recommended for routine filtration of aqueous solutions
  • Flasks have a hose connection for ¼" (6 mm) I. D. tubing
  • Very good chemical and temperature resistance
Recommended for large sample volumes, liquids with a heavy particulate load and viscous liquids when a high flow rate is required. Can provide filtration rates up to four times faster than 47 mm assemblies due to its larger filter area.
Glass components are manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité