Wheaton® Socorex™ Calibra® 832 Digital Macro Pipettes
Fournisseur: DWK Life Sciences
851345
WHEA851345EA 0 EUR InStock
WHEA851345
Wheaton® Socorex™ Calibra® 832 Digital Macro Pipettes
Pipettes Micropipettes
This Socorex® Calibra® 832 digital macropipette is a highly reliable macropipette with instant volume adjustment in a variable volume.
  • Single channel, tip color: natural
  • Calibra® 832 digital macropipette has a twin cam setting system with pre-calibrated steps that provide for true digital volume entry and numerical display
  • Plunger assembly features a stainless steel and PTFE construction for a high degree of chemical resistivity
  • Autoclavable, comes with a Three-year warranty
The Calibra® macropipette has a unique, permanently calibrated series of fixed stops, each one corresponding to one volume setting. This system provides accuracy, speed, and reliability.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité