Distillation glassware set with standard taper 19/22 Clear-Seal® joints, Micro Kit®, WHEATON®
Fournisseur: DWK Life Sciences
109933 123753 168353 964453 923769 773900 690170
WHEA109933EA 143 EUR InStock
WHEA109933 WHEA123753 WHEA168353 WHEA964453 WHEA923769 WHEA773900 WHEA690170
Distillation glassware set with standard taper 19/22 Clear-Seal® joints, Micro Kit®, WHEATON®
Composants de distillation Pièces pour distillation
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
  • Superior in vacuum and radioactive applications
  • Smooth surfaces resist contamination
  • Cleaning is simple and thorough
  • Interchangeable with ground joints
Apparatus for a wide range of organic chemistry experiments at a lower price than purchasing the components separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité