eShield™ Sterile Cell Phone Cover
Fournisseur: WHITNEY PRODUCTS INC
EC2100
WHIUEC2100EA 249 EUR InStock
WHIUEC2100
eShield™ Sterile Cell Phone Cover
Sachets Sacs de stockage
eShield™ sterile cell phone covers are designed to protect a sterile environment from contaminants on smart phones.
  • Covers are touchscreen compatible, even with double gloves
  • Phone covers easily accommodate aseptic technique
They are constructed of ultra-clear polyethylene film ensuring clear photos.
The sterile barrier provided by eShield™ covers enables the medical professional to use a smart phone to access studies, record data, conduct consultations, and capture photos conveniently and safely without contaminating the sterile environment.
They can be used within the sterile field in any medical environment, including surgical suites and clinical and research labs.
Certifications: FDA approved. Meets ASTM F1671 viral penetration guidelines.
Informations de commande: Contact your Avantor sales representative for more information.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité