HEATING MANTLES WHM, FOR 3000 ML REACTION VESSELS, CYLINDRICAL, A- 130 MM, HEIGHT 280 MM, INSIDE DEPTH: 210 MM, WITHOUT POWER CONTROLLER, TEMP. RANGE 450C, 550 W, ALUMINIUM CASING, POWER SUPPLY 230 V, SERIAL NO. 1 * 1 Pce
Fournisseur: WITEG LABORTECHNIK
HEATING MANTLES WHM, FOR 3000 ML REACTION VESSELS, CYLINDRICAL, A- 130 MM, HEIGHT 280 MM, INSIDE DEPTH: 210 MM, WITHOUT POWER CONTROLLER, TEMP. RANGE 450C, 550 W, ALUMINIUM CASING, POWER SUPPLY 230 V, SERIAL NO. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
HEATING MANTLES WHM, FOR 3000 ML REACTION VESSELS, CYLINDRICAL, A-Ø 130 MM, HEIGHT 280 MM, INSIDE DEPTH: 210 MM, WITHOUT POWER CONTROLLER, TEMP. RANGE 450°C, 550 W, ALUMINIUM CASING, POWER SUPPLY 230 V, SERIAL NO. 1 * 1 Pce WITGDH.WHM12214 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou