Heating mantles with reaction vessel, WiseTherm® WHM
Fournisseur: WITEG LABORTECHNIK GMBH
DH.WHM12216
WITGDH.WHM12216EA 329 EUR InStock
WITGDH.WHM12216
Heating mantles with reaction vessel, WiseTherm® WHM
Appareils chauffants Chauffe-ballons
Heating mantles with aluminium exterior and fabric interior. The integrated K-type thermo-sensor is basically embedded (except fabric mantles) which enables precise control of the mantles when used together with remote digital controller. External thermo-sensor is available, which enables precise control of the temperature of liquid sample in the vessel when used together with remote digital controller.
  • Maximum temperature 450 °C
  • Can be used with magnetic stirrers
  • Acid-resistant and robust standing
  • With coil type nickel chrome heating element
Informations de commande: Supplied without accessories, optional remote thermo controllers are available.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité