Incubating shaker, WIS-30
Fournisseur: WITEG LABORTECHNIK GMBH
DH.WIS00230
WITGDH.WIS00230EA 4010 EUR InStock
WITGDH.WIS00230
Incubating shaker, WIS-30
Agitateurs Agitateurs incubateurs
For applications in biological cultures at various temperatures up to 60 °C, this shaking incubator accommodates flasks up to 1000 ml. The front door and sliding platform enable easy loading and handling of samples. The unit features an advanced shaking mechanism, providing quiet shaking with precise speed control and temperature control by means of a digital fuzzy controller.
  • Digital back-lit LCD display
  • Universal spring rack for various shapes/size of flask available as an option
  • Automatic stop of shaking, heating and refrigeration when a door opens
  • RS232 interface
  • Timer: 99 h 59 min or continuous
Information de livraison: Supplied with WiseRemote® remote control software and cable. A range of options and accessories are available, please enquire for details.
Attention: Accommodates a maximum of 35×50/100, 16×200/300 or 12×500/1000 ml flasks.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité